missing translation for 'onlineSavingsMsg'
Learn More

Promega GloResponse™ Luciferase Reporter Cell Lines

Catalog No. PRE8520
Encompass
Change view
Click to view available options
Quantity:
2 Vials
Reporter Gene:
CRE-luc2P
NF-κB-luc2P
NFAT-RE-luc2P
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Reporter Gene
PRE8520 2 Vials NF-κB-luc2P
PRE8500 2 Vials CRE-luc2P
PRE8510 2 Vials NFAT-RE-luc2P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRE8520 Supplier Promega Supplier No. E8520

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Prebuilt, optimized luciferase reporter cell lines allow for the rapid development of a reporter assay based on the pathway of interest regulating the luciferase gene

  • Luciferase Reporter Cell Line with NF-κB Response Element
  • Rapid, bioluminescent detection of any cellular response resulting in modulation of NF-κB activities

Specifications

Content And Storage Below -140°C
Cell Line 293 (HEK)
Cell Type Kidney (Embryonic)
For Use With (Application) For rapid development of a reporter assay based on the pathway of interest regulating the luciferase gene
Quantity 2 Vials
Reporter Gene NF-κB-luc2P
Source Human Embryonic Kidney 293 Cells

By use of this product, researcher agrees to be bound by the terms of this limited use statement. If the researcher is not willing to accept the conditions of this limited use statement, and the product is unused, Promega will accept return of the unused product and provide the researcher with a full refund.

Researchers may use this product for research use only, no commercial use is allowed. Commercial Use means any and all uses of this product and derivatives by a party for monetary or other consideration and may include but is not limited to use in: (1) product manufacture; and (2) to provide a service, information or data; and/or resale of the product or its derivatives, whether or not such product or derivatives are resold for use in research. Researchers shall have no right to modify or otherwise create variations of the nucleotide sequence of the luciferase gene except that Researchers may: (1) create fused gene sequences provided that the coding sequence of the resulting luciferase gene has no more than four deoxynucleotides missing at the affected terminus compared to the intact luciferase gene sequence, and (2) insert and remove nucleic acid sequences in splicing research predicated on the inactivation or reconstitution of the luminescence of the encoded luciferase. No other use or transfer of this product or derivatives is authorized without the prior express written consent of Promega. In addition, Researchers must either: (1) use luminescent assay reagents purchased from Promega Corporation for all determinations of luminescence activity of this product and its derivatives; or (2) contact Promega to obtain a license for use of the product and its derivatives. Researchers may transfer derivatives to others for research use provided that at the time of transfer a copy of this label license is given to the recipients and recipients agree to be bound by the terms of this label license. With respect to any uses outside this label license, including any diagnostic, therapeutic or prophylactic uses, please contact Promega for supply and licensing information. Promega makes no representations or warranties of any kind, either expressed or implied, including for merchantability or fitness for a particular purpose with regards to the product. The terms of this agreement shall be governed under the laws of the State of Wisconsin, USA. The above license relates to Promega patents and/or patent applications on improvements to the luciferase gene.

  • U.S. Patent 5,670,356.
  • Australian Patent 2003272419 and other patents pending.
  • Commercial use of this cell line requires a license from AdVec Inc.
  • HEK293 cells were obtained under license from AdVec Inc.
  • The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713 and 5,700,673. A license (from Promega for research reagent products and from The Regents of the University of California for all other fields) is needed for any commercial sale of nucleic acid contained within or derived from this product.
  • Australian Patent 2001 285278 and other patents pending.
  • Mfr. No. E8510: The NFAT response element and its use are licensed under one or more of the following patents: U.S. Patents 5,837,840; 6,197,925; 5,989,810; 6,096,515; 6,388,052; 6,150,099; 6,171,781; 6,352,830; 6,312,899 and 6,875,571.