missing translation for 'onlineSavingsMsg'
Learn More

Promega GloMax™ 20/20 Luminometer: Replacement Tubing, Valves, Tips

Catalog No. PRE4851
Encompass
Change view
Click to view available options
Type:
Replacement Tubing, Valves and Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
PRE4851 Replacement Tubing, Valves and Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE4851 Supplier Promega Supplier No. E4851

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Two tubings, four valves, 30 tips

These services and accessories are intended for use with the GloMax 20/20 Luminometer.

  • For Use with GloMax(R) 20/20 Luminometer
  • Rental and Service Agreement also available

Specifications

Type Replacement Tubing, Valves and Tips
For Use With (Equipment) GloMax 20/20 Luminometer
Includes Two tubings, four valves, 30 tips
Storage Requirements 15 to 30°C