missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Flexi™ Vector System
Description
Includes
T4 DNA Ligase, Nuclease-free water, 5X Flexi digest buffer, 2X Flexi Ligase buffer, Flexi Enzyme blend (SgfI and PmeI)
- Directional Cloning of Protein-Coding Sequences
- Choose from a variety of initial applications (e.g., bacterial protein, mammalian or cell-free protein expression) and then transfer the insert as required
- Efficient insert transfer means direct use of recombinant clones with minimal time for screening background colonies
- Adaptable to high-throughput formats for large screening projects
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| For Use With (Application) | Clone ORF and conveniently transfer these inserts to a variety of vectors designed for multiple applications, directional cloning method for Protein-coding sequences, transfer Protein-coding regions without the need to resequence |
| Quantity | 100 Reactions |