Learn More
Promega ENLITEN™ ATP Assay System
The ENLITEN ATP Assay System is used to measure ATP levels for the indirect detection of biocontamination.
Supplier:  Promega FF2000
Encompass
Catalog No. PRFF2000
Only null left
Includes: 1 vial ENLITEN rLuciferase/Luciferin Reagent†; 12mL L/L Reconstitution Buffer; 1 vial ATP Standard; 25mL ATP-free water; 1 Protocol (TMF004)
Description
Description
  • Allows Indirect Detection of Biocontamination
Specifications
Specifications
100 Assays
-30°C to -10°C
Ideal for testing food processing surfaces, cosmetics and beverages and to assay Enzymes that degrade ATP or to quantitate ATP in biological fluids
1 x 1 vial Luciferase/Luciferin Reagent, 1 x 12mL L/L Reconstitution Buffer, 1 x 25mL ATP-free Water, 1 x 5mL ATP Standard
Product Suggestions
Product Suggestions