missing translation for 'onlineSavingsMsg'
Learn More

Promega dNTP Mix

A premixed solution containing sodium salts of dATP, dCTP, dGTP and dTTP, each at 10mM in water.

$51.39 - $109.36

Specifications

pH 7.5
Content And Storage -30°C to -10°C
Concentration 10 mM
Purity >99%
For Use With (Application) PCR amplification
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRU1515
Encompass
View Documents Promotion Details
Promega
U1515
1000 μL
Each for $109.36
Only null left
 
PRU1511
Encompass
View Documents Promotion Details
Promega
U1511
200 μL
Each for $51.39
Only null left
 
Description

Description

  • Ready to Use Solution of Deoxynucleotide Triphosphates
  • Highly pure (>99% triphosphate)
  • 10mM each nucleotide in water
  • Reduce pipetting steps by adding nucleotides as a mix
Specifications

Specifications

7.5
10 mM
PCR amplification
-30°C to -10°C
>99%
Videos
SDS
Documents

Documents