missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Promega™ DNA-Dependent Protein Kinase Peptide Substrate
Description
- Highly Specific Substrate for DNA-PK
- Supplied at 10mg/mL in water
- Km value for peptide is 760microM
Recommended Storage
−20°C
Patent pending. This product is covered by U.S. Patents 4,557,862 and 4,640,893 and is sold for research use only. All other uses, including, but not limited to, use as a clinical diagnostic or therapeutic, require a separate license. Please contact Prom
Specifications
Specifications
| Concentration | 10mg/mL |
| Formulation | 1 x 1mg DNA-dependent Protein Kinase Substrate |
| Molecular Weight (g/mol) | 1759.0 |
| pH Range | Neutral |
| Content And Storage | −30 to −10°C |
| Product Type | DNA-dependent Protein Kinase Peptide Substrate |
| Quantity | 1 mg |