missing translation for 'onlineSavingsMsg'
Learn More

Promega Digitonin Detergent Solution

Catalog No. PRG9441
Encompass
Change view
Click to view available options
Quantity:
40 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9441 40 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG9441 Supplier Promega Supplier No. G9441

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Digitonin is a detergent solution useful for permeabilizing cells and for creating a cytotoxicity chemistry positive control.

  • Detergent Solution for Permeabilizing Cells
  • Use as a cytotoxicity chemistry positive control
  • Supplied at 20mg/mL in DMSO

Specifications

Quantity 40 μL
Concentration 20 mg/mL
Content And Storage -30 to -10°C
For Use With (Application) Permeabilizing Cells