missing translation for 'onlineSavingsMsg'
Learn More

Promega Diamond™ Nucleic Acid Dye

Catalog No. PRH1181
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRH1181 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRH1181 Supplier Promega Supplier No. H1181

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Diamond Nucleic Acid Dye is a sensitive fluorescent dye that binds to single-stranded DNA, double-stranded DNA and RNA, and can be used to stain and visualize nucleic acids in gels.

  • Detect and visualize small amounts of nucleic acids after gel electrophoresis
  • Store at room temperature for quick and easy use without thawing
  • Use with a variety of common gel types and imaging equipment

Specifications

Boiling Point 189°C
Color Colorless
Form Liquid
For Use With (Application) Native Agarose, Polyacrylamide gels and standard imaging system
Quantity 500 μL
Storage Requirements −30 to −10°C
Product Type Fluorescent DNA/RNA Stain
Stain Type Fluorescent