Learn More
Promega Deoxyuridine Triphosphate (dUTP)
dUTP (2'-Deoxyuridine, 5'-Triphosphate) can be used in place of dTTP in PCR and RT-PCR to prevent carryover from previous amplifications.
Supplier: Promega U1191
Description
- High-Quality dNTPs, Including dUTP
- Highly pure (≥99% triphosphate)
- Each dNTP supplied at 100mM in water for ease-of-use
- Incorporate dUTP into PCR products and treat with uracil-N-glycosylase to inhibit subsequent amplifications
Chemical Identifiers
| 7.5 | |
| 100 mM | |
| cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols |
| -30°C to -10°C | |
| >99% |
Specifications
| 7.5 | |
| -30°C to -10°C | |
| >99% | |
| cDNA synthesis, sequencing and labeling, can be used in place of dTTP in PCR and RT-PCR protocols |
| dUTP | |
| 100 mM | |
| 40 μm |