missing translation for 'onlineSavingsMsg'
Learn More

Promega CytoTox-ONE™ Homogeneous Membrane Integrity Assay

Catalog No. PRG7891
Encompass
Change view
Click to view available options
Quantity:
1000 to 4000 Assays
200 to 800 Assays
Description:
Homogeneous Membrane Integrity Assay
Homogeneous Membrane Integrity Assay, HTP
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
PRG7891 1000 to 4000 Assays Homogeneous Membrane Integrity Assay
PRG7890 200 to 800 Assays Homogeneous Membrane Integrity Assay
PRG7892 1000 to 4000 Assays Homogeneous Membrane Integrity Assay, HTP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG7891 Supplier Promega Supplier No. G7891
Only null left

A fast homogeneous, fluorometric method for estimating the number of non-viable cells present in multiwell plates. Based on detection of LDH release, an indicator of cytotoxicity.

  • A Fluorometric Method for Estimating the Number of Nonviable Cells Present in Multiwell Plates
  • Elimination of cell transfer step saves time
  • Perform multiple assays on one sample

Specifications

Quantity 1000 to 4000 Assays
Description Homogeneous Membrane Integrity Assay
Format 96- or 384-well Plates
Content And Storage -30°C to -10°C
Product Line CytoTox-ONE
Detection Method Fluorometric
For Use With (Application) Cytotoxicity determination, total cell number determination, measures release of Lactate Dehydrogenase

U.S. Pat. Nos. 6,982,152 and 7,282,348, Australian Pat. No. 2003223521 and other patents pending.