missing translation for 'onlineSavingsMsg'
Learn More

Promega CytoTox-Fluor™ Cytotoxicity Assay

Catalog No. PRG9262
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 50 mL
5 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9262 2 x 50 mL
PRG9260 10 mL
PRG9261 5 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG9262 Supplier Promega Supplier No. G9262

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The CytoTox-Fluor Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the relative number of dead cells in cell populations.

  • A Fluorescent Assay to Measure the Relative Number of Dead Cells in Culture
  • Single-reagent-addition, homogeneous assay
  • Amenable to multiplexing with other luminescent cell-based assays

Specifications

Quantity 2 x 50 mL
Content And Storage -30°C to -10°C
Detection Method Fluorescence

Patent pending