Learn More
Promega CytoTox-Fluor™ Cytotoxicity Assay
The CytoTox-Fluor Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay that measures the relative number of dead cells in cell populations.
Supplier: Promega G9260
Description
- A Fluorescent Assay to Measure the Relative Number of Dead Cells in Culture
- Single-reagent-addition, homogeneous assay
- Amenable to multiplexing with other luminescent cell-based assays
Chemical Identifiers
| Fluorescence |
Specifications
| 10 mL | |
| Fluorescence | |
| -30°C to -10°C |
Patent pending