missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter-Fluor™ Cell Viability Assay

Catalog No. p-3604265
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 50 mL
5 x 10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6080 10 mL
PRG6081 5 x 10 mL
PRG6082 2 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG6080 Supplier Promega Supplier No. G6080
Only null left

The CellTiter-Fluor Cell Viability Assay is a non-lytic, single-reagent-addition fluorescence assay that measures the relative number of viable cells in a population.

  • A Non-Lytic Fluorescence Assay That Measures the Relative Number of Viable Cells in Culture
  • Multiplex with many other luminescent assays
  • Normalizing data for live-cell numbers make results more comparable to other data

Specifications

Quantity 10 mL
Detection Method Fluorescence
For Use With (Application) Normalizing Data for Live Cell Number
Kit Contents 1 x 10 μL GF-AFC substrate, 1 x 10 mL Assay Buffer
Format 96- or 384-well Plates
Content And Storage -30°C to -10°C

Patent Pending.