missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega CellTiter-Fluor™ Cell Viability Assay
Description
- A Non-Lytic Fluorescence Assay That Measures the Relative Number of Viable Cells in Culture
- Multiplex with many other luminescent assays
- Normalizing data for live-cell numbers make results more comparable to other data
Specifications
Specifications
| Quantity | 10 mL |
| Format | 96- or 384-well Plates |
| Content And Storage | 1 x 10 μL GF-AFC substrate, 1 x 10 mL Assay Buffer. Store at -30°C to -10°C |
| Detection Method | Fluorescence |
| For Use With (Application) | Normalizing Data for Live Cell Number |
Patent Pending.