missing translation for 'onlineSavingsMsg'
Learn More

Promega CellTiter 96™ Nonradioactive Cell Proliferation Assay (MTT)

Catalog No. PRG4100
Encompass
Change view
Click to view available options
Quantity:
1000 Assays
5000 Assays
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG4100 5000 Assays
PRG4000 1000 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG4100 Supplier Promega Supplier No. G4100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Colorimetric cell viability assay (MTT). Solubilization step is required.

  • A Colorimetric Assay to Assess Cell Viability
  • Greater sensitivity than the neutral red procedure
  • Non-radioactive, fast and convenient assay applicable to a variety of cells

Specifications

Quantity 5000 Assays
Format 96-well Plates
Content And Storage COMPONENTS AT DIFFERENT TEMPS
For Use With (Application) Cytotoxicity