missing translation for 'onlineSavingsMsg'
Learn More

Promega Cell Permeable Inhibitors

Catalog No. PRV1201
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1201 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV1201 Supplier Promega Supplier No. V1201

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

LY 294002 is a potent and specific cell-permeable inhibitor of phosphatidylinositol 3-kinases (PI 3-kinases).

  • Potent Inhibitor of Phosphatidylinositol 3-Kinases
  • Cell-permeable
  • Specifically inhibits PI 3-kinases
  • Supplied as a dried powder

Specifications

Quantity 5 mg
Content And Storage LY 294002. -30°C to -10°C
For Use With (Application) Blocks the activation of MEK1