Learn More
Promega cAMP-Glo™ Max Cyclic-AMP Assay DFS Item
The cAMP-Glo™ Max Assay is a homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells. Compounds that modulate GPCRs coupled with adenylate cyclase typically alter intracellular cAMP levels.
Supplier:  Promega V1682
 View more versions of this product
Catalog No. PRV1682
Only null left
Explore available promotions
Description
Description
  • Faster cAMP Detection for High-Throughput Assays
  • Complete in ∼30 minutes
  • Scalable to 1536-well plate formats and beyond
  • No interference by fluorescent compounds
Specifications
Specifications
-30°C to -10°C
500μL cAMP (1mM), 5mL MgCl2 (1M), 20mL cAMP-Glo ONE-buffer, 200μL protein Kinase A, 1 bottle Kinase-Glo substrate (lyophilized), 100mL Kinase-Glo buffer
Detection Assay Kit
A homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells
20 Preps
Product Suggestions
Product Suggestions