missing translation for 'onlineSavingsMsg'
Learn More

Promega cAMP-Glo™ Max Cyclic-AMP Assay

Catalog No. PRV1681
Encompass
Change view
Click to view available options
Quantity:
2 Preps
20 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1681 2 Preps
PRV1682 20 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1681 Supplier Promega Supplier No. V1681

The cAMP-Glo™ Max Assay is a homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells. Compounds that modulate GPCRs coupled with adenylate cyclase typically alter intracellular cAMP levels.

  • Faster cAMP Detection for High-Throughput Assays
  • Complete in ∼30 minutes
  • Scalable to 1536-well plate formats and beyond
  • No interference by fluorescent compounds

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) A homogeneous, bioluminescent and high-throughput assay to measure cyclic AMP (cAMP) levels in cells
Includes 100μL cAMP (1mM), 1mL MgCl2 (1M), 2.5mL cAMP-Glo ONE-buffer, 20μL protein Kinase A, 1 bottle Kinase-Glo substrate (lyophilized), 10mL Kinase-Glo buffer
Quantity 2 Preps
Type Detection Assay Kit