missing translation for 'onlineSavingsMsg'
Learn More

Promega BenchTop DNA Markers DFS Item

Ready-to-load markers offer the convenience of room temperature storage

$237.17 - $307.00

Specifications

Color Blue, Orange
Product Type DNA Molecular Weight Marker/Ladder
Form Liquid
Formulation Supplied in a stabilizing solution of 1X blue/orange loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
View More Specs

Products 5
Catalog Number Mfr. No. Quantity No. of Bands Concentration Price Qty  
Catalog Number Mfr. No. Quantity No. of Bands Concentration Price Qty  
PR-G7511 DFS Item
View Documents
Promega
G7511
250 μL 11 0.2 μg/μL N/A
Only null left
 
PR-G7521 DFS Item
View Documents
Promega
G7521
250 μL 15 0.2 μg/μL N/A
Only null left
 
PR-G7531 DFS Item
View Documents
Promega
G7531
300 μL 6 6.1 ng/μL N/A
Only null left
 
PR-G7541 DFS Item
View Documents
Promega
G7541
600 μL 13 0.08 μg/μL N/A
Only null left
 
PR-G8291 DFS Item
View Documents
Promega
G8291
300 μL 11 108 ng/μL N/A
Only null left
 
Description

Description

Supplied in a stabilizing solution of 1X Blue/Orange Loading Dye.

Specifications

Specifications

Blue, Orange
Liquid
Separation, size determination and quantitation of DNA and RNA fragments
DNA Molecular Weight Marker/Ladder
Supplied in a stabilizing solution of 1X blue/orange loading dye
−30 to +30°C
Videos
SDS
Documents

Documents