missing translation for 'onlineSavingsMsg'
Learn More

Promega AMV Reverse Transcriptase

Catalog No. PRM5101
Encompass
Change view
Click to view available options
Description:
AMV Reverse Transcriptase
AMV Reverse Transcriptase (HC)
Quantity:
300U
600U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
PRM5101 AMV Reverse Transcriptase 300U
PRM9004 AMV Reverse Transcriptase (HC) 600U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRM5101 Supplier Promega Supplier No. M5101

Includes

5X Reaction Buffer (250mM Tris-HCl [pH 8.3 at 25°C], 250mM KCl, 50mM MgCl2, 2.5mM spermidine, 50mM DTT)

AMV RT catalyzes DNA polymerization using template DNA, RNA or RNA:DNA hybrids.

  • Efficient Reverse Transcription of Templates with High Secondary Structure
  • DNA polymerization using template DNA, RNA or RNA:DNA hybrids
  • Requires a primer as well as magnesium or manganese
  • Stable at higher reaction temperatures (37-58°C)
  • Supplied at a concentration of 10u/μL (Cat.# M5101, M5108) or 20-25u/μL (Cat.# M9004)

Specifications

Concentration 10 U/μL
Content And Storage −30 to −10°C
Description AMV Reverse Transcriptase
Form Liquid
Quantity 300U
For Use With (Application) cDNA Synthesis
Includes 1 x 500μL 5X reaction buffer
Source Avian Myeloblastosis Virus
Storage Buffer Potassium Phosphate (pH 7.2 at °C) 200mM, 0.2% Triton™ X-100, DTT 2mM and Glycerol 50%