missing translation for 'onlineSavingsMsg'
Learn More

Promega 25bp DNA Step Ladder Molecular Weight Marker for Electrophoresis

Catalog No. PRG4511
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG4511 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG4511 Supplier Promega Supplier No. G4511

Includes

6X Blue/Orange Loading Dye

Twelve DNA fragments ranging from 25bp to 300bp in 25bp increments includes a visible 1,800bp backbone fragment.

  • Twelve DNA Fragments of 25-300bp in 25bp Increments
  • Includes 1,800bp backbone fragment
  • Load 5μL/lane
  • Supplied at a concentration of 0.36μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.4, 1mM EDTA
Color Blue, Orange
Concentration 0.36 μg/μL
Form Liquid
Formulation 100μg 25bp DNA Step ladder, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 12
Quantity 100 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker