missing translation for 'onlineSavingsMsg'
Learn More

Promega 200bp DNA Step Ladder Molecular Weight Marker for Electrophoresis

Catalog No. PRG6961
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6961 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG6961 Supplier Promega Supplier No. G6961
Only null left

Includes

6X Blue/Orange Loading Dye

Thirty-three blunt-ended DNA fragments ranging from 200bp to 6,600bp in 200bp increments. The 1,000bp band appears more intense than all other bands.

  • Thirty-Three Blunt-Ended DNA Fragments of 200-6,600bp in 200bp Increments
  • Load 1μL/lane
  • Supplied at a concentration of 1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.4, 1mM EDTA
Color Blue, Orange
Concentration 1 μg/μL
Form Liquid
Formulation 100μg 200bp DNA Step ladder, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 33
Quantity 100 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker