missing translation for 'onlineSavingsMsg'
Learn More

Promega 1kb DNA Step Ladder Molecular Weight Marker for Electrophoresis

Catalog No. PRG6941
Encompass
Change view
Click to view available options
Quantity:
90 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6941 90 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG6941 Supplier Promega Supplier No. G6941
Only null left

Includes

6X Blue/Orange Loading Dye

Ten blunt-ended DNA fragments ranging from 1kb to 10kb in 1kb increments.

  • Ten Blunt-Ended DNA Fragments of 1-10kb in 1kb Increments
  • Load 1μL/lane
  • Supplied at a concentration of 0.3μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.4, 1mM EDTA
Color Blue, Orange
Concentration 300 ng/μL
Form Liquid
Formulation 90μg 1kb DNA Step ladder, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 10
Quantity 90 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker