missing translation for 'onlineSavingsMsg'
Learn More

Promega 1kb DNA Ladder Molecular Weight Marker

Catalog No. PRG5711
Encompass
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG5711 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG5711 Supplier Promega Supplier No. G5711
Only null left

Includes

6X Blue/Orange Loading Dye

13 blunt-end fragments from 250bp to 10,000bp.

  • Thirteen Blunt-Ended Fragments of 250-10,000bp
  • Load 5μL/lane
  • Supplied at a concentration of 0.1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Color Blue, Orange
Concentration 0.1 μg/μL
Form Liquid
For Use With (Application) Determining the size of DNA fragment
Kit Contents 1 × 500μL 1kb DNA ladder, 1 × 1mL blue/orange 6X loading dye
No. of Bands 13
Quantity 500 μL
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker