missing translation for 'onlineSavingsMsg'
Learn More

Promega 10bp DNA Step Ladder Molecular Weight Marker for Electrophoresis

Catalog No. PRG4471
Encompass
Change view
Click to view available options
Quantity:
32.5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG4471 32.5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG4471 Supplier Promega Supplier No. G4471
Only null left

Includes

6X Blue/Orange Loading Dye

Ten blunt-ended DNA fragments ranging from 10bp to 100bp in exactly 10bp increments

  • Ten Blunt-Ended DNA Fragments of 10-100bp in 10bp Increments
  • Load 1μL/lane
  • Supplied at a concentration of 0.65μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.4, 1mM EDTA
Color Blue, Orange
Concentration 0.65 μg/μL
Form Liquid
Formulation 50μL 10bp DNA Step ladder, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 10
Quantity 32.5 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker