missing translation for 'onlineSavingsMsg'
Learn More

Promega 100bp DNA Step Ladder Molecular Weight Marker for Electrophoresis

Catalog No. PRG6951
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG6951 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRG6951 Supplier Promega Supplier No. G6951
Only null left

Includes

6X Blue/Orange Loading Dye

Forty blunt-ended DNA fragments ranging from 100bp to 4,000bp in 100bp increments.

  • Forty Blunt-Ended DNA fragments of 100-4,000bp in 100bp Increments
  • Load 1μL/lane
  • Supplied at a concentration of 1μg/μL
  • Supplied with 6X Blue/Orange Loading Dye

Specifications

Buffer 10mM Tris HCl pH 7.4, 1mM EDTA
Color Blue, Orange
Concentration 1 μg/μL
Form Liquid
Formulation 100μg 100bp DNA Step ladder, 1mL blue/orange 6X loading dye
For Use With (Application) Separation, size determination and quantitation of DNA and RNA fragments
No. of Bands 40
Quantity 100 μg
Storage Requirements −30 to −10°C
Product Type DNA Molecular Weight Marker