missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProLong™ Glass Antifade Mountant with NucBlue™ Stain

Catalog No. P36985
Encompass
Change view
Click to view available options
Quantity:
2 mL
10 mL
5 x 2 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P36985 10 mL
P36983 2 mL
P36981 5 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P36985 Supplier Invitrogen™ Supplier No. P36985

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ProLong Glass Antifade with NucBlue Stain contains the nuclear counterstain Hoechst 33342 to detect DNA and provides sample and fluorescence preservation. ProLong Glass exhibits the same RI as glass and immersion oil (1.52) for high resolution microscopy imaging of cell and tissue samples up to 150 microns in thickness.

ProLong Glass Antifade with NucBlue Stain is a ready-to-use hard-setting mounting media with a refractive index of 1.52 for high resolution microscopy imaging that contains the nuclear counterstain Hoechst 33342 to detect DNA. ProLong Glass Antifade Mountant with NucBlue Stain can be applied directly to fluorescently labeled cells or tissue samples on slides or coverslips. When bound to DNA, NucBlue (Hoechst 33342) is excited by UV light at 360 nm with an emission maximum at 460 nm and is detected using a DAPI traditional filter. ProLong Glass mountant has a refractive index (RI) of 1.52, similar to glass coverslips, compatible immersion oil, and oil-immersion microscope optics, which enables superior resolution and sensitivity. ProLong Glass mounting media is designed to provide unparalleled photobleach protection across the visible and near infra-red spectra. You can use ProLong Glass Antifade Mountant with NucBlue Stain with almost any fluorescent dye or fluorescent protein (e.g., GFP, RFP, mCherry) and with any cell/tissue sample up to 150 microns in thickness. With embedded nuclear DNA counterstain of NucBlue within the ProLong Glass mounting media, multiplexing is smooth and effortless.

ProLong Glass Antifade Mountant with NucBlue Stain is provided in a 2 mL dropper bottle or 10 mL bottle. After removing excess water, just simply apply a drop of mounting media to the sample, add a coverslip, cure, and image. ProLong Glass Mountant with NucBlue Stain minimizes optical refraction and spherical aberration in the optical path, which enables longer-term storage of your samples. ProLong Glass with NucBlue Stain mountant does not discolor or shrink when cured and stored long term and provides nuclear counterstaining directly in the mounting media for DNA detection.

Optimal refractive index for the brightest and sharpest images
The RI determines how much the path of light is bent, or refracted, when entering a material. In microscopy, refractive index differences between specimen and mounting medium should be minimized and can be reduced by using mountants with a higher RI such as ProLong Glass mountant. Depending on your sample type, thickness, imaging depth and/or staining reagents (e.g. fluorescent proteins or dyes) and whether you need a curing or non-curing mountant, choosing the right mounting media will impact image resolution and should be strongly considered for optimal experimental design. For example, when Prolong Glass Antifade Mountant (1.52 RI) is compared with a mounting medium with a refractive index of 1.47, the axial resolution is improved by 32% at a focal depth of 40 μm and 75% at a focal depth of 100 μm. The signal intensity and sensitivity of fluorophores is also improved as the focal depth increases within the sample. Prolong Glass is optimal for acquisition of Z-Stack, 3D, and 2D images of cells and tissue up to 150 μm while providing superior nuclear labeling of most cells and tissues alongside NucBlue stain to assess DNA with nuclear staining.

Prevent photobleaching across the visible and near-IR spectra
Compatible with almost all fluorescent dyes and fluorescent proteins, ProLong Glass Antifade Mountant with NucBlue provides superior protection against photobleaching. ProLong Glass mounting media does not quench fluorophores and provides high-resolution images with a wide array of fluorescent dyes, including Alexa Fluor, FITC, TRTC, Texas Red, Cy3, and Cy5 dyes while NucBlue for nuclear DNA staining is delivered within the mountant. ProLong Glass Antifade Mountant is an ideal reagent for archiving samples and for preserving signal strength during prolonged imaging sessions, such as repeated scanning under confocal laser scanning illumination or with super-resolution microscopy,

Glass mountants are recommended for all immersion oil imaging applications and for samples greater than 10 microns. ProLong Glass Antifade Mountant with NucBlue stain contains an optimized concentration of Hoechst 33342 stain, which provides superior nuclear labeling of DNA within most cells and tissues. However, specimens thicker than 30 μm can require optimization and longer incubations. For specimens thicker than 30 μm, consider pre-incubation with labeling reagents such as NucBlue Live ReadyProbes Reagent followed by mounting with ProLong Glass Antifade Mountant. To image your sample immediately after NucBlue staining, use our SlowFade Glass Antifade Mountant, a non-curing mountant with similar 1.52 refractive index properties.

Order Info

Store at 2–8°C. For long-term storage, store at -5 to -30°C. Avoid freeze/thaw cycles.

Specifications

Quantity 10 mL
Product Type Antifade Mountant
Content And Storage Store at 2°C to 8°C.
Shipping Condition Approved for shipment on Wet or Dry Ice
Product Line ProLong
Reagent Type Antifade Solution
Volume (Metric) 10 mL
Description ProLong™ Glass Antifade Mountant with NucBlue™ Stain, 1 x 10 mL
I mounted my antibody-labeled brain cryosection in either ProLong Antifade Mountant, ProLong Gold, ProLong Diamond or ProLong Glass Antifade Mountant. It looked good when first mounted, but after curing, it had tiny bubbles all over the tissue under the coverslip. The sample was air-dried prior to mounting. What happened and what can be done to remove the bubbles?

The tissue likely had air trapped in it, either before the labeling process or as a result of air-drying. While the tissue is in blocking solution or another wash, we recommend putting it in a vacuum to pull out internalized air. Air-drying is not necessary; just tap off excess buffer and mount the damp tissue with the mountant.

I do not have epoxy or VALAP to seal the coverslip. Do you offer an alternative coverslip sealant?

We offer ProLong Coverslip Sealant (Cat. No. P56128) that can be used to seal the edges of the coverslip and is compatible with both curing and non-curing mountant. The sealant is easy to apply and is brushed on after the mountant has cured, for long preservation of slides. The product page can be found here.

When using a hard-curing mountant, such as ProLong Antifade Mountant, make sure the mountant is fully cured before applying ProLong Coverslip Sealant. Since this sealant can seal moisture under the coverslip, it can interfere with the curing process.

When using a non-curing mountant, such as SlowFade Antifade Mountants, sealing can take place immediately after mounting.

What do ProLong Gold, ProLong Diamond and ProLong Glass antifade mountant reagents contain?

They are composed of proprietary antifades and polymers in aqueous buffers.

Can I use ProLong Gold, ProLong Diamond, or ProLong Glass mounting medium in a multi-well plate?

No. If the sample cannot be sealed with a coverslip, we recommend the use of SlowFade antifade mountant.

What can affect sample curing of ProLong Gold-, ProLong Diamond-, and ProLong Glass-mounted samples?

Our ProLong Gold, ProLong Diamond, and ProLong Glass mounting media harden ('cure') by the evaporation of water. Placing the mounted samples in a humid or refrigerated environment would slow down the evaporation process ('curing').

What is the concentration of NucBlue Stain in ProLong Glass Antifade Mountant with NucBlue Stain?
The concentration of NucBlue dye in this product is proprietary.

Are ProLong, ProLong Gold, ProLong Diamond, and ProLong Glass reagents compatible with DyLight dye-conjugated antibodies and streptavidin and with other organic dyes?

We have not tested ProLong, ProLong Gold, ProLong Diamond, or ProLong Glass on DyLight dye-conjugated proteins or with other organic dyes, but it is likely that they will be compatible.

Can I use any of the ProLong, ProLong Gold, ProLong Diamond, or ProLong Glass reagents on live samples?

No. ProLong, ProLong Gold, ProLong Diamond, and ProLong Glass reagents are intended for use on fixed or fixed/permeabilized samples. Only ProLong Live Antifade Reagent (Cat Nos. P36974 and P36975) is intended for use with live cells.

What happens when ProLong, ProLong Gold, ProLong Diamond, or ProLong Glass antifade mountant is curing? Is there a chemical reaction?

The evaporation of water is the “curing” process for Prolong, ProLong Gold, ProLong Diamond, and ProLong Glass antifade mountant. The evaporation of water causes the polymer to harden. There is no chemical reaction. Please note that there is no curing/hardening for any of the SlowFade reagents or ProLong Live products.

How do I remove bubbles from the vials of either ProLong Antifade Mountant, ProLong Gold, ProLong Diamond or ProLong Glass Antifade Mountants or ProLong-mounted samples?

Bubbles may be removed by one of two methods:

1. Place the amount of ProLong reagent you wish to use on your sample (plus a little excess) in a microcentrifuge tube. Close the cap and centrifuge this aliquot using a tabletop microcentrifuge (speed from 7, 000 to 13,000 rpm). Bubbles should move to the top and these bubbles may be aspirated using a pipettor/pipette tip.
2. Unscrew the lid of the bottle/vial containing the ProLong reagent to make it loose, but do not remove the lid. Place the entire bottle/vial into a vacuum flask, using a faucet aspirator (faucet T-tube). Apply a vacuum (water running through the faucet) and allow vacuum aspiration to occur from 10 to 20 minutes to degas the mixture.

To avoid the formation of bubbles on a sample or to remove bubbles:

1. Before pipetting the desired amount of ProLong reagent for mounting, set the pipettor for a slight excess volume. When pipetting up the mixture, do not pipette up the complete amount, but lift up the pipette tip from the bottle with the pipettor not yet up to full volume. This prevents the aspiration of bubbles into the pipette tip.
2. Bubbles trapped during application of the coverslip: When placing your coverslip onto your drop of ProLong reagent, place the coverslip at a slight angle then, gently lower the coverslip. If the coverslip is lowered flat onto the sample, or lowered too quickly, bubbles can be trapped.
3. Bubbles trapped in tissue: One problem with tissue sections, particularly cryosections is that air can get trapped within and under the section. Upon mounting, bubbles are not observed but as the mountant hardens, it compresses the sample slightly, forcing air out of the section. This leads to microscopic bubbles forming over the section, trapped within the mountant. To avoid this, degas the tissue sample prior to mounting. Place the sections submerged in buffer or blocking solution, into a vacuum chamber and expose the sample to the vacuum. This will degas the sections and buffer. Remove the sample from this degassed buffer and mount.
4. If ProLong-mounted samples have already cured but have bubbles, you can un-mount your sample by placing the slides into PBS (Coplin jar or a Petri dish filled with PBS). The ProLong reagent will swell and the coverslip will slide off or can be gently removed manually. You can then re-mount with a new aliquot of ProLong reagent.


For Research Use Only. Not for use in diagnostic procedures.