missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STERIS ProKlenz™ Sterile Detergent
A concentrated detergent for use in controlled environments
Specifications
| pH | Neutral |
|---|---|
| Sterility | Sterile |
| Filtered | Filtered |
| For Use With (Application) | Removing Organic/Inorganic Residues From Product or Non-Product Contact Surfaces |
Description
- Designed to remove organic and inorganic residues from product and non-product contact surfaces
- Formulated with multiple surfactants to clean a broad spectrum of soils
- Neutral pH allows for use in manual cleaning and compatibility with many substrates
- Filtered and filled in an ISO Class 5 aseptic environment to minimize the potential of particles being introduced during use
- Packages are double bagged for easy entry into controlled environments and aseptic cores
- Includes an extensive documentation package to meet validation objectives
- Each lot documented by a Certificate of Analysis that includes sterility results
- Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
| Neutral | |
| Sterile | |
| Filtered | |
| Removing Organic/Inorganic Residues From Product or Non-Product Contact Surfaces |