missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ ProKlenz RESTORE High Performance Acid Based Cleaner

Catalog No. 17103045
Change view
Click to view available options
Size:
5 gal.
55 gal.
Container Type:
Drum
Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17103045 5 gal. Pail
17103046 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17103045 Supplier STERIS™ Supplier No. 142305WR

ProKlenz RESTORE is a globally available, low-foaming acid-based product for type I rouge removal and all-purpose cleaning. It effectively removes corrosion (rouge) at reduced concentrations, lower temperatures, and shorter contact times compared to other acid-based detergents.

ProKlenz RESTORE High Performance Acid Based Cleaner is a globally available, low foaming acid based product for type I rouge removal and all-purpose cleaning. The global acceptability of ProKlenz RESTORE accommodates the pharmaceutical, biotechnology, cosmetic, and nutraceutical industries by removing difficult process residues.

HOW PROKLENZ RESTORE WORKS

ProKlenz RESTORE Cleaner has an acidic pH for the removal of type I rouge and all-purpose cleaning.

WHY PROKLENZ RESTORE?

  • Standardization - Globally available product enables harmonized cleaning protocols across sites.
  • Sustainability - Free rinsing formula minimizes utility consumption and detergent residue
  • Effectiveness - An acidic pH removes type I rouge while a low foaming system supports use in high-impingement systems
  • Efficiency - A low use-concentration supports cost-effective and optimized cleaning
  • Quality - Manufactured within an FDA registered plant, ISO 9001:2015 and ISO 13485:2016 compliant, to meet the highest standard in manufacturing and quality assurance

Specifications

Product Type Acid Cleaner
Color-coded No
Form Clear Liquid
pH 2.4 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Pail
Size 5 gal.