Learn More
STERIS™ ProKlenz ALIGN Neutral Pharmaceutical Detergent
ProKlenz ALIGN Neutral Detergent offers gentle but effective cleaning for a wide range of process residues in CIP and manual applications. ProKlenz ALIGN Detergent has broad substrate compatibility and a neutral pH that eliminates the need for wastewater neutralization.
$481.32
Specifications
| Product Type | Detergent |
|---|---|
| Color-coded | No |
| Form | Clear Liquid |
| pH | 8.4 (Diluted) |
| Scent | Unscented |
Description
ProKlenz ALIGN Neutral Pharmaceutical Detergent is a globally available, low foaming neutral-based product for cleaning. ProKlenz ALIGN effectively cleans difficult residues in the pharmaceutical, biotechnology, and oral solid dose industries.
HOW PROKLENZ ALIGN WORKS
ProKlenz ALIGN Neutral Pharmaceutical Detergent is engineered to clean a wide range of soils and materials with a neutral pH. This product is low-foaming for clean-in-place or manual cleaning applications. ProKlenz ALIGN is a neutral detergent to promote excellent substrate compatibility, water effluent discharge requirements, and operator safety.
WHY PROKLENZ ALIGN?
- Productivity - Effective cleaning against a wide variety of pharmaceutical process residues in clean-in-place (CIP), clean-out-of-place (COP) and manual applications.
- Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports.
- Efficacy - Neutral pH meets the requirements for operator safety in manual cleaning and water effluent restrictions.
- Globally Availability - Manufactured with biodegradable ingredients to support harmonization of cleaning processes.
- Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility.
Specifications
| Detergent | |
| Clear Liquid | |
| Unscented |
| No | |
| 8.4 (Diluted) | |
| Non-sterile |