missing translation for 'onlineSavingsMsg'
Learn More

STERIS™ ProKlenz 120 High Performance Alkaline Detergent

Catalog No. p-200076303
Change view
Click to view available options
Size:
55 gal.
Container Type:
Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Container Type
17183984 55 gal. Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17183984 Supplier STERIS™ Supplier No. 143201WR
Only null left

ProKlenz 120 Mild Alkaline Detergent is formulated to meet the unique cleaning challenges found in the pharmaceutical and other industries. ProKlenz 120 Detergent’s low foaming and free rinsing formula can be used in high-energy spray applications or manual cleaning. pH (1%): 11.5, typical.

ProKlenz™ 120 Alkaline Detergent is specially formulated to deliver effective cleaning with excellent substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

How ProKlenz 120 Works:

ProKlenz 120 Alkaline Detergent is a phosphate-free, liquid, alkaline detergent for use in clean-in-place (CIP), clean-out-of-place (COP), and manual cleaning applications. It is formulated for enhanced materials compatibility.

pH (1%): 11.5, typical

Why ProKlenz 120?

  • Formulated alkaline detergent for effective removal of residues in both soft and hard water
  • Formulated for excellent materials compatibility
  • Reduces utility consumption and cleaning time with low foaming, free-rinsing formulation
  • Phosphate-free formulation with biodegradable surfactants
  • Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility.

Specifications

Product Type Detergent
Color-coded No
Form Clear Liquid
pH 11.5 (Diluted)
Scent Unscented
Sterility Non-sterile
Container Type Drum
Size 55 gal.