Learn More
MilliporeSigma™ Progard™ TL1-C Pretreatment Pack
Catalog No. PR0GTLCS1
Change view
Click to view available options
Type:
Water Purification Consumables
Catalog No. Type
PR0GTLCS1 Water Purification Consumables
1 options
Catalog No. PR0GTLCS1 Supplier MilliporeSigma™ Supplier No. PR0GTLCS1
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services.
  • Designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories
  • Can be used in both validated and non-validated environments
  • The Progard™ Pretreatment Pack has been designed to pretrat water by removing large particles, organic compounds and free chlorine from tap water
  • This protects the reverse osmosis (RO) membrane against mineral scaling, organic fouling and chlorine oxidation
  • Benefits:
    • Optimal protection of reverse osmosis (RO) membrane
    • Ensures system lifetime and performance are optimized
Specifications
Type Water Purification Consumables
For Use With (Application) For optimal protection of Reverse Osmosis (RO) membranes
Includes ROProtect C for automatic RO maintenance