Learn More
MilliporeSigma™ Proguard™ TS2 Pretreatment Pack
For optimal protection of reverse osmosis membranes. Proguard™ TS2 Pretreatment Packs are designed to remove large particles and organic compounds, softening water, and removing chlorine from tap water.
Supplier:  MilliporeSigma™ PR0G0T0S2
Catalog No. PR0G0T0S2
Only null left
Explore available promotions
Description
Description
For optimal protection of reverse osmosis membranes. Proguard™ TS2 Pretreatment Packs are designed to remove large particles and organic compounds, softening water, and removing chlorine from tap water.
  • Activated carbon
  • Anti-scaling
  • Depth filtration
Specifications
Specifications
Activated Carbon, Anti-scaling, Depth Filtration
Optimal Protection of Reverse Osmosis Membranes (with Bacteriostatic Activated Carbon)
1 Each
Pretreatment Pack
Product Suggestions
Product Suggestions