missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 816 Thermal Fingerless Winter Work Gloves/Flip-Top Mittens

Catalog No. 19820157
Change view
Click to view available options
Size:
Small
Medium
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820157 Small
19820158 Medium
19820160 X-Large
19820161 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19820157 Supplier Ergodyne™ Supplier No. 17342
Only null left

Mittens feature a soft microfleece lining for cold-weather protection and easily convert to half-finger gloves for detail work.

  • Mittens convert to half-finger gloves for detailed work
  • Flip-top mitten and thumb stow easily in mitten pocket when not in use
  • Soft microfleece lining provides comfort, flexibility, and protection in cold conditions
  • Rugged synthetic leather palm maintains flexibility in cold temps
  • Reinforced thumb saddle extends glove life
  • Contoured gathered elastic cuff with ID space
  • Gloves fit over jacket sleeves for convenient on/off wear
  • Reflective accents enhance wearer visibility and hand signaling
  • Soft thumb nose/brow wipe area

Specifications

Color Black
Hand Specific
Material Polyester
Size Small
EN388 Standard 3423 X
For Use With (Application) Construction, Cold Weather Detail Work, Cold Storage/Refrigeration, and Outdoor Recreation
Glove Cuff Style Elasticated
Liner Material Microfleece
Palm Type Synthetic Leather
Series ProFlex™ 816