missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 260 Soft Slip-On Knee Pads

Catalog No. 19820275
Change view
Click to view available options
slip-on knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820275 slip-on knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820275 Supplier Ergodyne™ Supplier No. 18260

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Economical light duty slip-on knee protection for a range of applications.

Specifications

Color Black
For Use With (Application) Flooring, Landscaping, Maintenance,Carpentry, Mechanics, Trades
Liner Material Cloth/Foam