missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 821 Smooth Surface Handling Work Gloves

Catalog No. 19820116
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820116 Small
19820117 Medium
19820118 Large
19820119 X-Large
19820120 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820116 Supplier Ergodyne™ Supplier No. 17232

Palm and index fingers feature a silicone coating to provide a secure grip when handling smooth surfaces such as boxes or cartons.

  • Breathable polyester mesh keeps hands cool without being bulky or stiff
  • Low-profile, molded hook & loop closure provides a secure fit
  • Gloves feature an ID tab for personalization to prevent misplaced gloves
  • Pull-on tab for easy on/off
  • Machine washable

Specifications

Coating Material Silicone
Color Black
Hand Specific
Material Polyester Mesh
Product Type Work Gloves
Size Small
For Use With (Application) Warehousing/Distribution, Package Delivery
Glove Cuff Style Open
Series ProFlex™ 821