missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 335HL Slip Resistant Rubber Cap Knee Pads

Catalog No. 19820279
Change view
Click to view available options
slip resistant knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820279 slip resistant knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820279 Supplier Ergodyne™ Supplier No. 18336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Grips wet or dry work surfaces. Hook and loop closure.

Specifications

Color Black Cap
For Use With (Application) Construction, Masonry, Freight/Baggage,Flooring, Maintenance, Utilities, Carpentry, Guerilla Warfare
Material 840D Nylon