missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 4000 Single Strap Wrist Support

Catalog No. 19035272F
Change view
Click to view available options
Color:
Black
Tan
Size:
Small
Medium
X-Large
Hand:
Left
Right
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size Hand
19035272F Black X-Large Left
19035274C Tan Small Left
19035274D Tan Medium Left
19910438 Tan X-Large Left
19035273F Tan X-Large Right
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19035272F Supplier Ergodyne™ Supplier No. 70018

Single strap support with open center supports the wrist during repetitive motion discomfort by limiting wrist movements.

Specifications

Color Black
Size X-Large
For Use With (Application) Food Processing,Picking/Packing,Assembly/Fabrication
Hand Left