missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 325HL Non-Marring Rubber Cap Knee Pads

Catalog No. 19820278
Change view
Click to view available options
non-marring knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820278 non-marring knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820278 Supplier Ergodyne™ Supplier No. 18326

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Will not damage or mark delicate work surfaces.

Specifications

Color Tan Cap
For Use With (Application) Flooring, Maintenance, Utilities,Construction, Masonry, Freight/Baggage,Carpentry
Material 840D Nylon