missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 315HL Long Textured Hard Cap Knee Pads

Catalog No. 19820277
Change view
Click to view available options
hard cap knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820277 hard cap knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820277 Supplier Ergodyne™ Supplier No. 18316

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Stands up to tough environments.

Specifications

Color White Cap
For Use With (Application) Construction, Masonry, Freight/Baggage,Flooring, Maintenance, Utilities, Carpentry
Material 840D Nylon