missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 230LTR Leather Knee Pads - Wide Soft Cap

Catalog No. 19820274
Change view
Click to view available options
leather knee pads
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19820274 leather knee pads
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820274 Supplier Ergodyne™ Supplier No. 18232

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Built tough with real leather to provide excellent traction and knee protection from fire, slag and sparks.

  • Leather is abrasion-resistant from the toughest work surfaces
  • Pads wrap over the top of the knee pad for extra durability and a longer life for both knees and the knee pads, even for jobs that require working on the knees daily.
  • Made with nitrile rubber (NBR) foam padding that keeps its shape over time and helps avoid common knee injuries.
  • Built slightly wider than traditional knee pads, for added coverage and comfort
  • Non-slip dual straps easily adjust to most knee/leg sizes
  • Bckle closure keep the knee pad secured in place all day long without restricting circulation
  • Anti-odor treatment protects the inner fabric to keep pads fresh
  • Ideal for construction, flooring and concrete applications
  • Resistance to heat, fire and abrasion makes for great welding knee pads

Specifications

Color Brown
For Use With (Application) Construction,Assembly,Fabrication,Pulp and Paper,Service Technicians
Liner Material NBR foam