missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 7141 Hi-Vis Nitrile-Coated Cut-Resistant Gloves

Catalog No. p-7163186
Change view
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large
Catalog No. Size
19820258 Small
19820259 Medium
19820260 Large
19820261 X-Large
19820262 2X-Large
5 options

Catalog No. 19820258

Supplier: Ergodyne™ 17912

Only null left

Designed with two new technologies to create comfortable, breathable and secure grip ANSI cut level 4 gloves. Features molded TPR Armor™ on the knuckles and fingers for dorsal impact protection.

  • Made with high-performance polyethylene (HPPE) fibers for A4 cut protection
  • HPPE does not include steel or glass often used in other anti-cut gloves which can cause irritation
  • WSX™ dip combined with TenaLux™ yarn make these palm-coated gloves cool to the touch with 360-degrees of breathability
  • Lighter weight than traditional A4 cut-resistant gloves
  • Hi-vis lime color on the backside of the hand
  • Liquid-resistant special coating absorbs and wicks moisture from the surface allowing for a firm-grip in wet oily and wet conditions
  • Water-based polyurethane (PU) coating combined with nitrile dip allows maximum dexterity and significantly relieves hand fatigue while ensuring durability
  • Molded TPR Armor™ on the backside of the glove provides pinch injury prevention and blow defection
  • TPR™ extends to fingertips and is sewn on for maximum durability
  • WearArmor™ flex zones deliver protection without sacrificing dexterity or flexibility
  • Ideal for workers in heavy construction, oil and gas, mining, demolition, extraction and refining, ironwork or cargo handling

Specifications

ANSI Abrasion Rating ANSI 4
Construction Seamless Knit
Coating Material Nitrile
Color Lime Green, Black
Cut Resistance Rating ANSI A4
Material HPPE
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Construction, assembly, fabrication, pulp and paper, service technicians
Grip Design WSX dip technology
Model 17912
Series ProFlex™ 7141
Show More Show Less