missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ergodyne™ ProFlex™ 710CR Heavy-Duty Cut-Resistant Gloves
Description
- ANSI A4 cut protection with Armortex-backed palms
- Tena-Grip padded palm, finger and thumb saddle reinforcements for use with wet or dry materials
- Comfortable, technical hex-stretch knit is not bulky or stiff
- Neoprene knuckle pad for increased back-of-hand protection
- Low-profile, molded hook and loop closure
- Machine washable
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 388: 3544 D |
| Color | Gray |
| Cut Resistance Rating | ANSI A4 |
| Product Type | Cut-resistant Gloves |
| Size | Large |
| For Use With (Application) | Heavy Duty Work,Construction,Assembly/Fabrication,Sheet Metal,Sharp Edge Material Handling,Electrical,Pulp and Paper,Mechanics,Service Techs,Tool/Machine Operation |