missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 4010 Double Strap Wrist Support

Catalog No. 19035277E
Change view
Click to view available options
Hand:
Right
Color:
Tan
Size:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Hand Color Size
19035277E Right Tan Large
19035277D Right Tan Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19035277E Supplier Ergodyne™ Supplier No. 70126

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Double strap wrist support with open center supports the wrist during repetitive motion discomfort by limiting wrist movements.

Specifications

Color Tan
Size Large
For Use With (Application) Food Processing, Picking/Packing,Assembly/Fabrication
Hand Right