missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 925WP Performance Dorsal Impact-Reducing Thermal Waterproof Gloves

Catalog No. p-7163144
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820268 Small
19820270 Large
19820271 X-Large
19820272 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19820268 Supplier Ergodyne™ Supplier No. 18102
Only null left

Gloves combine 3M™ Thinsulate™ Insulation and Full TPR Armor to reduce the effects of impacts during cold applications. Breathable membrane insert is waterproof and windproof for long lasting protection.

  • Dual-zone 3M™ Thinsulate™ Insulation
  • Waterproof/windproof breathable membrane insert
  • Full TPR armor with technical foam for maximum protection
  • Dual-layer AX Suede™-reinforced gripping palm
  • Armortex™-reinforced thumb saddle and index finger
  • Padded neoprene cuff with reflective binding
  • Pull-on tab with ID space for personalization

Specifications

Back Material TPR Armor
Cuff Material Neoprene
Color Black
Hand Specific
Product Type Anti-impact Gloves
Size Small
EN388 Standard EN388:3323 XP
For Use With (Application) Cold Weather Work, Oil/Gas, Mining, and Heavy Construction
Glove Cuff Style Open
Liner Material 3M™ Thinsulate™
Palm Type Dual-Layer AX Suede™
Series ProFlex™ 925WP
Show More Show Less