missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 925F(x) Standard Dorsal Impact-Reducing Gloves

Catalog No. 19820257
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820257 2X-Large
19820253 Small
19820254 Medium
19820255 Large
19820256 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820257 Supplier Ergodyne™ Supplier No. 17906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gloves are constructed of full thermoplastic rubber armor with technical foam padding for additional protection. Palms feature Hi-Vis AX Suede™ non-slip dots for enhanced gripping capabilities.

  • Full TPR armor with technical foam padding
  • Hi-Vis AX Suede™ with non-slip dot gripping palm
  • Armortex™-reinforced thumb saddle and index finger
  • Hi-vis breathable mesh construction
  • Padded neoprene cuff with reflective binding
  • Pull-on tab with ID space to write user's name

Specifications

Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size 2X-Large
For Use With (Application) Oil/Gas Drilling, Mining, and Heavy Construction
Glove Cuff Style Open
Series ProFlex™ 925F(x)