missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 924LTR Leather-Reinforced Hybrid Dorsal Impact-Reducing Gloves

Catalog No. 19820240
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820240 Small
19820241 Medium
19820242 Large
19820243 X-Large
19820245 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820240 Supplier Ergodyne™ Supplier No. 17792

Gloves feature thermoplastic rubber construction throughout the fingers with a fully padded, genuine leather reinforced palm.

  • Leather-reinforced fingers and thumb saddle
  • Hi-vis breathable mesh construction
  • Neoprene cuff with molded hook and loop closure keeps out debris
  • Gloves feature ID space to write user's names

Specifications

Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Small
For Use With (Application) Roofing, Mining, Oil/Gas Drilling, Construction, and Mechanics
Glove Cuff Style Hook and Loop
Series ProFlex™ 924LTR