missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 924 Hybrid Dorsal Impact-Reducing Gloves

Catalog No. p-7163140
Change view
Click to view available options
Size:
Small
Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820193 Small
19820195 Large
19820197 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19820193 Supplier Ergodyne™ Supplier No. 17682
Only null left

Gloves feature thermoplastic rubber construction throughout the fingers, with flexible foam knuckle padding for additional impact protection.

  • Padded palm reinforced with Tena-Grip™ technology
  • Hi-vis breathable mesh construction
  • TPR fingertips with foam knuckle padding and a reinforced thumb saddle
  • Neoprene cuff with molded hook and loop closure keeps out debris
  • Gloves feature ID space to write user's name

Specifications

Back Material Mesh
Cuff Material Neoprene
Color Hi-vis Lime Green, Black
Hand Specific
Material Thermoplastic Rubber
Product Type Impact-Reducing Gloves
Size Small
For Use With (Application) Oil/Gas Drilling, Mining, Construction, and Mechanics
Glove Cuff Style Hook and Loop
Series ProFlex™ 924