missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 910 Half-Finger Impact Gloves with Wrist Support

Catalog No. 19820211
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820211 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19820211 Supplier Ergodyne™ Supplier No. 17714

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AVC™ padding in the palm and other key areas helps to reduce shock and impact. Built-in wrist support with Open-Center Stay™ technology relieves discomfort caused by repetitive motion.

  • Segmented AVC™ palm padding reduces shock and palm impact
  • Coated palm provides enhanced grip capabilites
  • Built-in wrist support with patented Open-Center Stay™ aides in relief from repetitive motion discomfort
  • Half-finger design for maximum dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Neoprene knuckle pad with reinforced thumb saddle
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Cuff Material Neoprene
Color Black
Hand Specific
Material Spandex
Product Type Anti-impact Gloves
Size Large
For Use With (Application) Construction, Manufacturing, Repetative Motion Tasks, Hand and Power Tools
Glove Cuff Color Black
Glove Cuff Style Hook and Loop
Series ProFlex™ 910