missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ ProFlex™ 901 Half-Finger Leather Impact Gloves

Catalog No. 19820239
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19820239 2X-Large
19820235 Small
19820236 Medium
19820237 Large
19820238 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19820239 Supplier Ergodyne™ Supplier No. 17786

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Foam padding in the palm and other key areas helps to reduce shock and impact. genuine leather construction provides a natural, yet durable feel.

  • Segmented foam palm padding reduces shock and impact
  • Genuine leather palm provides durability with a natural feel
  • Half-finger design for dexterity and tactile feedback
  • Hex stretch knit and breathable spandex construction
  • Finger webbing loops for easy on/off
  • Neoprene knuckle pad
  • Low-profile molded hook & loop closure
  • Gloves feature ID space to write user's name

Specifications

Back Material Spandex
Color Black
Hand Specific
Material Geniune Leather
Product Type Anti-impact Gloves
Size 2X-Large
For Use With (Application) Construction, Manufacturing, Hand and Power Tools
Glove Cuff Style Hook and Loop
Series ProFlex™ 901